missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-fluorobenzylamine hydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC290510050

 View more versions of this product

Catalog No. AC290510050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
240.5
Yellow
77771-03-0
BrC6H3(F)CH2NH2·HCl
97.5
MFCD00143094
Authentic
Powder
5g
100.0
C7H7BrFN·HCl
98%
3-Bromo-4-fluorobenzylamine hydrochloride
98%
Glass bottle

Chemical Identifiers

77771-03-0
100.0
C7H7BrFN·HCl
240.5
3-Bromo-4-fluorobenzylamine hydrochloride, 98%
97.5
98%
BrC6H3(F)CH2NH2·HCl
MFCD00143094
≥97.5%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA