Please login to your online account to display your discounted pricing

3-Bromo-4-fluorobenzylamine hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-fluorobenzylamine hydrochloride
Name Note 98%
CAS 77771-03-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7BrFN·HCl
Linear Formula BrC6H3(F)CH2NH2·HCl
Formula Weight 240.5
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC290510050

  SDS
thermo fisher scientific
AC290510050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 240.5
Quantity 5g
Color Yellow
TSCA TSCA
CAS Max % 100.0
CAS 77771-03-0
Molecular Formula C7H7BrFN·HCl
Linear Formula BrC6H3(F)CH2NH2·HCl
Name Note 98%
CAS Min % 97.5
Chemical Name or Material 3-Bromo-4-fluorobenzylamine hydrochloride
MDL Number MFCD00143094
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass bottle