missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-fluorobenzotrifluoride, TCI America™

$51.95 - $375.84

Chemical Identifiers

CAS 68322-84-9
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 242.999
MDL Number MFCD00040938
InChI Key RZJOIMPUMMQKFR-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorobenzotrifluoride, 2-bromo-1-fluoro-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethyl bromobenzene, 3-bromo-alpha,alpha,alpha,4-tetrafluorotoluene, benzene, 2-bromo-1-fluoro-4-trifluoromethyl, 1-bromo-2-fluoro-5-trifluoromethyl benzene, 3-bromo-alpha,alpha,alpha-4-tetrafluorotoluene, pubchem1639, 3-bromo-4-fluorobenzotrifluoride, 3-bromo-4-fluoro trifluoromethyl benzene
PubChem CID 144295
IUPAC Name 2-bromo-1-fluoro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Br)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B14211G View Documents TCI America
B14211G
1g Each for $51.95
 
B142125G View Documents TCI America
B142125G
25g Each for $375.84
 
B14215G View Documents TCI America
B14215G
5g Each for $125.28
 
Specifications

Chemical Identifiers

68322-84-9
242.999
RZJOIMPUMMQKFR-UHFFFAOYSA-N
144295
C1=CC(=C(C=C1C(F)(F)F)Br)F
C7H3BrF4
MFCD00040938
3-bromo-4-fluorobenzotrifluoride, 2-bromo-1-fluoro-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethyl bromobenzene, 3-bromo-alpha,alpha,alpha,4-tetrafluorotoluene, benzene, 2-bromo-1-fluoro-4-trifluoromethyl, 1-bromo-2-fluoro-5-trifluoromethyl benzene, 3-bromo-alpha,alpha,alpha-4-tetrafluorotoluene, pubchem1639, 3-bromo-4-fluorobenzotrifluoride, 3-bromo-4-fluoro trifluoromethyl benzene
2-bromo-1-fluoro-4-(trifluoromethyl)benzene

Specifications

60°C
1g
C7H3BrF4
3-bromo-4-fluorobenzotrifluoride, 2-bromo-1-fluoro-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethyl bromobenzene, 3-bromo-alpha,alpha,alpha,4-tetrafluorotoluene, benzene, 2-bromo-1-fluoro-4-trifluoromethyl, 1-bromo-2-fluoro-5-trifluoromethyl benzene, 3-bromo-alpha,alpha,alpha-4-tetrafluorotoluene, pubchem1639, 3-bromo-4-fluorobenzotrifluoride, 3-bromo-4-fluoro trifluoromethyl benzene
C1=CC(=C(C=C1C(F)(F)F)Br)F
242.999
243.00
≥96.0% (GC)
Yellow
68322-84-9
MFCD00040938
RZJOIMPUMMQKFR-UHFFFAOYSA-N
2-bromo-1-fluoro-4-(trifluoromethyl)benzene
144295
Clear Liquid at 20°C
3-Bromo-4-fluorobenzotrifluoride
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator