missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-fluorobenzotrifluoride, 97%

$38.18 - $561.84

Chemical Identifiers

CAS 68322-84-9
Molecular Formula C7H3BrF4
Formula Weight 243
MDL Number MFCD00040938
Beilstein 2093911
Synonym 2-Bromo-1-fluoro-4-(trifluoromethyl)benzene; 2-Fluoro-5-(trifluoromethyl)bromobenzene
Chemical Name or Material 3-Bromo-4-fluorobenzotrifluoride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2007803 SDS Alfa Aesar™
B2007803
1g Each for $38.18
AAB2007806 SDS Alfa Aesar™
B2007806
5g Each for $142.69
AAB2007814 SDS Alfa Aesar™
B2007814
25g Each for $561.84
Specifications Description & Specifications

Chemical Identifiers

68322-84-9
243
2093911
3-Bromo-4-fluorobenzotrifluoride
C7H3BrF4
MFCD00040938
2-Bromo-1-fluoro-4-(trifluoromethyl)benzene; 2-Fluoro-5-(trifluoromethyl)bromobenzene

Specifications

68322-84-9
243
2093911
148° to 149°C
71°C (159°F)
97%
C7H3BrF4
MFCD00040938
2-Bromo-1-fluoro-4-(trifluoromethyl)benzene; 2-Fluoro-5-(trifluoromethyl)bromobenzene
1g
3-Bromo-4-fluorobenzotrifluoride

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning