missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-cyanopyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 13958-98-0
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.01
InChI Key VPDIXJCBBUZNPO-UHFFFAOYSA-N
Synonym 3-bromo-4-cyanopyridine, 3-bromo-4-pyridinecarbonitrile, 3-bromo-isonicotinonitrile, 3-bromoisonicotinonitrile, 4-pyridinecarbonitrile, 3-bromo, 4-pyridinecarbonitrile,3-bromo, 5-bromoisonicotinonitrile, acmc-20a0fj, pubchem17105
PubChem CID 11074051
IUPAC Name 3-bromopyridine-4-carbonitrile
SMILES C1=CN=CC(=C1C#N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC452270010
SDS View Documents
Thermo Fisher Scientific
AC452270010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC452270050
SDS View Documents
Thermo Fisher Scientific
AC452270050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

13958-98-0
183.01
3-bromo-4-cyanopyridine, 3-bromo-4-pyridinecarbonitrile, 3-bromo-isonicotinonitrile, 3-bromoisonicotinonitrile, 4-pyridinecarbonitrile, 3-bromo, 4-pyridinecarbonitrile,3-bromo, 5-bromoisonicotinonitrile, acmc-20a0fj, pubchem17105
3-bromopyridine-4-carbonitrile
C6H3BrN2
VPDIXJCBBUZNPO-UHFFFAOYSA-N
11074051
C1=CN=CC(=C1C#N)Br

Specifications

91°C to 94°C
13958-98-0
C6H3BrN2
VPDIXJCBBUZNPO-UHFFFAOYSA-N
3-bromopyridine-4-carbonitrile
11074051
97%
1g
96% min. (GC)
3-bromo-4-cyanopyridine, 3-bromo-4-pyridinecarbonitrile, 3-bromo-isonicotinonitrile, 3-bromoisonicotinonitrile, 4-pyridinecarbonitrile, 3-bromo, 4-pyridinecarbonitrile,3-bromo, 5-bromoisonicotinonitrile, acmc-20a0fj, pubchem17105
C1=CN=CC(=C1C#N)Br
183.01
183.01
3-Bromo-4-cyanopyridine
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS Signal Word: Danger

SDS