missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-4-cyanopyridine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC452270010
Specifications
3-Bromo-4-cyanopyridine | |
1g | |
96% min. (GC) | |
3-bromo-4-cyanopyridine, 3-bromo-4-pyridinecarbonitrile, 3-bromo-isonicotinonitrile, 3-bromoisonicotinonitrile, 4-pyridinecarbonitrile, 3-bromo, 4-pyridinecarbonitrile,3-bromo, 5-bromoisonicotinonitrile, acmc-20a0fj, pubchem17105 | |
C1=CN=CC(=C1C#N)Br | |
183.01 | |
183.01 |
91°C to 94°C | |
13958-98-0 | |
C6H3BrN2 | |
VPDIXJCBBUZNPO-UHFFFAOYSA-N | |
3-bromopyridine-4-carbonitrile | |
11074051 | |
97% |
Chemical Identifiers
13958-98-0 | |
183.01 | |
3-bromo-4-cyanopyridine, 3-bromo-4-pyridinecarbonitrile, 3-bromo-isonicotinonitrile, 3-bromoisonicotinonitrile, 4-pyridinecarbonitrile, 3-bromo, 4-pyridinecarbonitrile,3-bromo, 5-bromoisonicotinonitrile, acmc-20a0fj, pubchem17105 | |
3-bromopyridine-4-carbonitrile |
C6H3BrN2 | |
VPDIXJCBBUZNPO-UHFFFAOYSA-N | |
11074051 | |
C1=CN=CC(=C1C#N)Br |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
GHS Signal Word: Danger