Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-cyanopyridine, 95%

$302.86 - $690.09

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-cyanopyridine
CAS 13958-98-0
Molecular Formula C6H3BrN2
Formula Weight 183.01
MDL Number MFCD00234143
Synonym 3-Bromoisonicotinonitrile; 3-Bromopyridine-4-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2704903

  SDS
alfa aesar™
H2704903
1g Each for $690.09

AAH27049MD

  SDS
alfa aesar™
H27049MD
250mg Each for $302.86
Description & Specifications

Specifications

Formula Weight 183.01
Chemical Name or Material 3-Bromo-4-cyanopyridine
Assay 95%
Molecular Formula C6H3BrN2
MDL Number MFCD00234143
CAS 13958-98-0
Synonym 3-Bromoisonicotinonitrile; 3-Bromopyridine-4-carbonitrile
Melting Point 92° to 94°C
Quantity 1g
Odor Odorless
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.