missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-chloropentafluorobutyric acid, 97%

$42.55 - $495.03

Chemical Identifiers

CAS 216393-99-6
Molecular Formula C4HBrClF5O2
Molecular Weight (g/mol) 291.396
MDL Number MFCD00153071
InChI Key IFQBWLXWJWDSTE-UHFFFAOYSA-N
Synonym 3-bromo-4-chloropentafluorobutyric acid, acmc-1cmca, 3-bromo-4-chloro-2,2,3,4,4-pentafluorobutyric acid, butanoic acid,3-bromo-4-chloro-2,2,3,4,4-pentafluoro, 3-bromanyl-4-chloranyl-2,2,3,4,4-pentakis fluoranyl butanoic acid
PubChem CID 2736232
IUPAC Name 3-bromo-4-chloro-2,2,3,4,4-pentafluorobutanoic acid
SMILES C(=O)(C(C(C(F)(F)Cl)(F)Br)(F)F)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1671103 View Documents Alfa Aesar™
L1671103
1g Each for $42.55
 
AAL1671106 View Documents Alfa Aesar™
L1671106
5g Each for $135.70
 
AAL1671114 View Documents Alfa Aesar™
L1671114
25g Each for $495.03
 
Specifications

Chemical Identifiers

216393-99-6
291.396
IFQBWLXWJWDSTE-UHFFFAOYSA-N
2736232
C(=O)(C(C(C(F)(F)Cl)(F)Br)(F)F)O
C4HBrClF5O2
MFCD00153071
3-bromo-4-chloropentafluorobutyric acid, acmc-1cmca, 3-bromo-4-chloro-2,2,3,4,4-pentafluorobutyric acid, butanoic acid,3-bromo-4-chloro-2,2,3,4,4-pentafluoro, 3-bromanyl-4-chloranyl-2,2,3,4,4-pentakis fluoranyl butanoic acid
3-bromo-4-chloro-2,2,3,4,4-pentafluorobutanoic acid

Specifications

57°C (0.1mmHg)
216393-99-6
MFCD00153071
3-bromo-4-chloropentafluorobutyric acid, acmc-1cmca, 3-bromo-4-chloro-2,2,3,4,4-pentafluorobutyric acid, butanoic acid,3-bromo-4-chloro-2,2,3,4,4-pentafluoro, 3-bromanyl-4-chloranyl-2,2,3,4,4-pentakis fluoranyl butanoic acid
C(=O)(C(C(C(F)(F)Cl)(F)Br)(F)F)O
291.396
291.4
3-Bromo-4-chloropentafluorobutyric acid
1g
C4HBrClF5O2
UN3265
IFQBWLXWJWDSTE-UHFFFAOYSA-N
3-bromo-4-chloro-2,2,3,4,4-pentafluorobutanoic acid
2736232
97%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.