missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4-chloronitrobenzene, TCI America™

$59.58 - $174.17

Chemical Identifiers

CAS 16588-26-4
Molecular Formula C6H3BrClNO2
Molecular Weight (g/mol) 236.449
MDL Number MFCD00100437
InChI Key CGTVUAQWGSZCFH-UHFFFAOYSA-N
PubChem CID 2735549
IUPAC Name 2-bromo-1-chloro-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Br)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B16245G View Documents TCI America
B16245G
5g Each for $59.58
 
B162425G View Documents TCI America
B162425G
25g Each for $174.17
 
Specifications

Chemical Identifiers

16588-26-4
236.449
CGTVUAQWGSZCFH-UHFFFAOYSA-N
2-bromo-1-chloro-4-nitrobenzene
C6H3BrClNO2
MFCD00100437
2735549
C1=CC(=C(C=C1[N+](=O)[O-])Br)Cl

Specifications

Yellow
5g
C6H3BrClNO2
CGTVUAQWGSZCFH-UHFFFAOYSA-N
2-bromo-1-chloro-4-nitrobenzene
2735549
Crystal-Powder at 20°C
3-Bromo-4-chloronitrobenzene
58°C
16588-26-4
MFCD00100437
C1=CC(=C(C=C1[N+](=O)[O-])Br)Cl
236.449
236.45
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No