missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-4-chloronitrobenzene 97.0+%, TCI America™
Supplier: TCI America B162425G
Specifications
3-Bromo-4-chloronitrobenzene | |
58°C | |
C6H3BrClNO2 | |
25 g | |
[O-][N+](=O)C1=CC(Br)=C(Cl)C=C1 | |
236.45 | |
236.45 | |
Crystalline Powder |
16588-26-4 | |
Yellow | |
MFCD00100437 | |
CGTVUAQWGSZCFH-UHFFFAOYSA-N | |
2-bromo-1-chloro-4-nitrobenzene | |
2735549 | |
≥97.0% (GC) |
Chemical Identifiers
16588-26-4 | |
236.45 | |
CGTVUAQWGSZCFH-UHFFFAOYSA-N | |
2-bromo-1-chloro-4-nitrobenzene |
C6H3BrClNO2 | |
MFCD00100437 | |
2735549 | |
[O-][N+](=O)C1=CC(Br)=C(Cl)C=C1 |
Safety and Handling
TSCA : No