Learn More
3-Bromo-4-chlorobenzotrifluoride, 98+%, ACROS Organics™
Chemical Identifiers
CAS | 454-78-4 |
---|---|
Molecular Formula | C7H3BrClF3 |
Molecular Weight (g/mol) | 259.45 |
MDL Number | MFCD00018093 |
InChI Key | APSISOSWYXCEQX-UHFFFAOYSA-N |
Synonym | 2-bromo-1-chloro-4-trifluoromethyl benzene, 3-bromo-4-chloro-a,a,a-trifluorotoluene, 3-bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene, 3-bromo-4-chloro-benzotrifluoride, 3-bromo-4-chlorobenzotrifluoride, 4-chloro-3-bromobenzotrifluoride, apsisoswyxceqx-uhfffaoysa, benzene, 2-bromo-1-chloro-4-trifluoromethyl, ksc915a8j, pubchem4837 |
PubChem CID | 67988 |
IUPAC Name | 2-bromo-1-chloro-4-(trifluoromethyl)benzene |
SMILES | C1=CC(=C(C=C1C(F)(F)F)Br)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC137510050
|
Thermo Fisher Scientific
137510050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC137510250
|
Thermo Fisher Scientific
137510250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
454-78-4 | |
259.45 | |
APSISOSWYXCEQX-UHFFFAOYSA-N | |
67988 | |
C1=CC(=C(C=C1C(F)(F)F)Br)Cl |
C7H3BrClF3 | |
MFCD00018093 | |
2-bromo-1-chloro-4-trifluoromethyl benzene, 3-bromo-4-chloro-a,a,a-trifluorotoluene, 3-bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene, 3-bromo-4-chloro-benzotrifluoride, 3-bromo-4-chlorobenzotrifluoride, 4-chloro-3-bromobenzotrifluoride, apsisoswyxceqx-uhfffaoysa, benzene, 2-bromo-1-chloro-4-trifluoromethyl, ksc915a8j, pubchem4837 | |
2-bromo-1-chloro-4-(trifluoromethyl)benzene |
Specifications
1.7400g/mL | |
Authentic | |
1.4980 to 1.5 | |
188°C to 190°C | |
-23°C to 22°C | |
454-78-4 | |
100.0 | |
C7H3BrClF3 | |
MFCD00018093 | |
APSISOSWYXCEQX-UHFFFAOYSA-N | |
2-bromo-1-chloro-4-(trifluoromethyl)benzene | |
67988 | |
Liquid | |
3-Bromo-4-chlorobenzotrifluoride, 98+% |
94°C | |
Glass bottle | |
1.74 | |
Yellow | |
5g | |
98.0 | |
98+% | |
BrC6H3(Cl)CF3 | |
2-bromo-1-chloro-4-trifluoromethyl benzene, 3-bromo-4-chloro-a,a,a-trifluorotoluene, 3-bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene, 3-bromo-4-chloro-benzotrifluoride, 3-bromo-4-chlorobenzotrifluoride, 4-chloro-3-bromobenzotrifluoride, apsisoswyxceqx-uhfffaoysa, benzene, 2-bromo-1-chloro-4-trifluoromethyl, ksc915a8j, pubchem4837 | |
C1=CC(=C(C=C1C(F)(F)F)Br)Cl | |
259.45 | |
259.45 | |
98+% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 207-226-0
TSCA : TSCA