Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-4-chlorobenzotrifluoride, 97%

$29.70 - $29.70

Chemical Identifiers

Chemical Name or Material 3-Bromo-4-chlorobenzotrifluoride
CAS 454-78-4
Molecular Formula C7H3BrClF3
Formula Weight 259.44
MDL Number MFCD00018093
Beilstein 1638470
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2366606

 
alfa aesar™
B2366606
5g Each for $29.70
Description & Specifications

Specifications

Formula Weight 259.44
Chemical Name or Material 3-Bromo-4-chlorobenzotrifluoride
Assay 97%
Molecular Formula C7H3BrClF3
MDL Number MFCD00018093
CAS 454-78-4
Melting Point −23° to −22°C
Beilstein 1638470
Boiling Point 191° to 192°C
Quantity 5g
Flash Point 94°C (201°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.