missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-4,5-dimethoxybenzaldehyde, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC450090010
Specifications
3-Bromo-4, 5-dimethoxybenzaldehyde | |
61°C to 63°C | |
6948-30-7 | |
C9H9BrO3 | |
Solubility in water: insoluble | |
COC1=C(C(=CC(=C1)C=O)Br)OC | |
245.07 | |
245.07 |
Glass bottle | |
1g | |
96% min. (GC) | |
3,4-dimethoxy-5-bromobenzaldehyde, 3-bromo-4,5-dimethoxy benzaldehyde, 5-bromoveratraldehyde, 5-bromoveratric aldehyde, 5-bromveratrumaldehyde, acmc-209ml7, benzaldehyde, 3-bromo-4,5-dimethoxy, benzaldehyde,3-bromo-4,5-dimethoxy, cambridge id 7187634 | |
ICVODPFGWCUVJC-UHFFFAOYSA-N | |
3-bromo-4,5-dimethoxybenzaldehyde | |
81379 | |
97% |
Chemical Identifiers
6948-30-7 | |
245.07 | |
3,4-dimethoxy-5-bromobenzaldehyde, 3-bromo-4,5-dimethoxy benzaldehyde, 5-bromoveratraldehyde, 5-bromoveratric aldehyde, 5-bromveratrumaldehyde, acmc-209ml7, benzaldehyde, 3-bromo-4,5-dimethoxy, benzaldehyde,3-bromo-4,5-dimethoxy, cambridge id 7187634 | |
3-bromo-4,5-dimethoxybenzaldehyde |
C9H9BrO3 | |
ICVODPFGWCUVJC-UHFFFAOYSA-N | |
81379 | |
COC1=C(C(=CC(=C1)C=O)Br)OC |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 230-113-2