missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-4,5-dimethoxybenzaldehyde, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450090050

 View more versions of this product

Catalog No. AC450090050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Bromo-4, 5-dimethoxybenzaldehyde
61°C to 63°C
6948-30-7
C9H9BrO3
Solubility in water: insoluble
COC1=C(C(=CC(=C1)C=O)Br)OC
245.07
245.07
Glass bottle
5g
96% min. (GC)
3,4-dimethoxy-5-bromobenzaldehyde, 3-bromo-4,5-dimethoxy benzaldehyde, 5-bromoveratraldehyde, 5-bromoveratric aldehyde, 5-bromveratrumaldehyde, acmc-209ml7, benzaldehyde, 3-bromo-4,5-dimethoxy, benzaldehyde,3-bromo-4,5-dimethoxy, cambridge id 7187634
ICVODPFGWCUVJC-UHFFFAOYSA-N
3-bromo-4,5-dimethoxybenzaldehyde
81379
97%

Chemical Identifiers

6948-30-7
245.07
3,4-dimethoxy-5-bromobenzaldehyde, 3-bromo-4,5-dimethoxy benzaldehyde, 5-bromoveratraldehyde, 5-bromoveratric aldehyde, 5-bromveratrumaldehyde, acmc-209ml7, benzaldehyde, 3-bromo-4,5-dimethoxy, benzaldehyde,3-bromo-4,5-dimethoxy, cambridge id 7187634
3-bromo-4,5-dimethoxybenzaldehyde
C9H9BrO3
ICVODPFGWCUVJC-UHFFFAOYSA-N
81379
COC1=C(C(=CC(=C1)C=O)Br)OC
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 230-113-2

SDS Documents