Please login to your online account to display your discounted pricing

3-Bromo-2-nitrotoluene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-nitrotoluene
CAS 52414-97-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H6BrNO2
Formula Weight 216.05
MDL Number MFCD02673631
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC357850100

  SDS
thermo fisher scientific
AC357850100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Low Melting Mass
Assay ≥97.5%
Formula Weight 216.05
Quantity 10g
Color Yellow
Melting Point 26° to 30°C
CAS Max % 100.0
Chemical Name or Material 3-Bromo-2-nitrotoluene
Assay Percent Range 98%
Molecular Formula C7H6BrNO2
MDL Number MFCD02673631
CAS 52414-97-8
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Infrared Spectrum Authentic