missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-nitroaniline, 98%

Manufacturer:  Alfa Aesar™ H6116906

 View more versions of this product

Catalog No. AAH6116906



Specifications

Specifications

3-Bromo-2-nitroaniline
5g
C6H5BrN2O2
UN2811
GLKHLBAXHLAQBJ-UHFFFAOYSA-N
3-bromo-2-nitroaniline
21852072
Crystalline
Red
7138-15-0
MFCD08063248
3-bromo-2-nitrobenzenamine, 2-amino-6-bromonitrobenzene, 3-bromo-2-nitrophenylamine, 3-bromo-2-nitro-phenylamine, pubchem3139, acmc-209oig, 3-bromo-2-nitro-aniline, ksc376s4f, 3-amino-2-nitrobromobenzene
C1=CC(=C(C(=C1)Br)[N+](=O)[O-])N
217.022
217.02
98%

Chemical Identifiers

7138-15-0
217.022
GLKHLBAXHLAQBJ-UHFFFAOYSA-N
21852072
C1=CC(=C(C(=C1)Br)[N+](=O)[O-])N
C6H5BrN2O2
MFCD08063248
3-bromo-2-nitrobenzenamine, 2-amino-6-bromonitrobenzene, 3-bromo-2-nitrophenylamine, 3-bromo-2-nitro-phenylamine, pubchem3139, acmc-209oig, 3-bromo-2-nitro-aniline, ksc376s4f, 3-amino-2-nitrobromobenzene
3-bromo-2-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents