Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-methylpyridine, 97%

$123.55 - $319.43

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-methylpyridine
CAS 38749-79-0
Molecular Formula C6H6BrN
Formula Weight 172.03
MDL Number MFCD00191224
Synonym 3-Bromo-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2727103

  SDS
alfa aesar™
H2727103
1g Each for $319.43

AAH27271MD

  SDS
alfa aesar™
H27271MD
250mg Each for $123.55
Description & Specifications

Specifications

Formula Weight 172.03
Boiling Point 76°C (17mmHg)
Quantity 1g
Chemical Name or Material 3-Bromo-2-methylpyridine
Assay 97%
CAS 38749-79-0
Health Hazard 3 GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 3-Bromo-2-picoline
Health Hazard 2 GHS H Statement
H301-H311-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C6H6BrN
MDL Number MFCD00191224
Flash Point 79°C (174°F)
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.