Learn More
3-Bromo-2-methylpropene, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 427650050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Bromo-2-methylpropene | |
96.0 | |
1.3390g/mL | |
7°C | |
Glass bottle | |
CH2=C(CH3)CH2Br | |
5mL | |
3-bromo-2-methylpropene, 3-bromo-2-methyl-1-propene, methallyl bromide, 2-bromomethyl prop-1-ene, 2-methylallyl bromide, 1-propene, 3-bromo-2-methyl, 3-bromo-2-methyl-prop-1-ene, 3-bromo-2-methylpropene, stabilized with hydroquinone, methallylbromide, methylallyl bromide | |
CC(=C)CBr | |
135.00 | |
135 |
1458-98-6 | |
100.0 | |
94.0°C to 95.0°C | |
96% min. (GC) | |
C4H7Br | |
MFCD00134155 | |
1.339 | |
USEGQJLHQSTGHW-UHFFFAOYSA-N | |
3-bromo-2-methylprop-1-ene | |
357785 | |
97% |
Chemical Identifiers
1458-98-6 | |
135.00 | |
USEGQJLHQSTGHW-UHFFFAOYSA-N | |
357785 | |
CC(=C)CBr |
C4H7Br | |
MFCD00134155 | |
3-bromo-2-methylpropene, 3-bromo-2-methyl-1-propene, methallyl bromide, 2-bromomethyl prop-1-ene, 2-methylallyl bromide, 1-propene, 3-bromo-2-methyl, 3-bromo-2-methyl-prop-1-ene, 3-bromo-2-methylpropene, stabilized with hydroquinone, methallylbromide, methylallyl bromide | |
3-bromo-2-methylprop-1-ene |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away fr
GHS Signal Word: Danger
RUO â Research Use Only