Please login to your online account to display your discounted pricing

3-Bromo-2-methylimidazo[1,2-a]pyridine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC21710DA

 View more versions of this product

Catalog No. CC21710DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 211.06
Quantity 1g
Assay 97%
Molecular Formula C8H7BrN2
Packaging Amber Glass Bottle
Assay Percent Range 97%
Chemical Name or Material 3-Bromo-2-methylimidazo[1, 2-a]pyridine
CAS 4805-70-3

Specifications

Chemical Name or Material 3-Bromo-2-methylimidazo[1, 2-a]pyridine
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H7BrN2
Formula Weight 211.06