Please login to your online account to display your discounted pricing

3-Bromo-2-methylimidazo[1,2-a]pyridine, 97%, Maybridge

$201.23 - $201.23

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-methylimidazo[1, 2-a]pyridine
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H7BrN2
Formula Weight 211.06
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC21710DA

  SDS
fisher scientific company
CC21710DA
Amber Glass Bottle 1g Each for $201.23

CC21710DE

 
fisher scientific company
CC21710DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 211.06
Quantity 1g
Assay 97%
Molecular Formula C8H7BrN2
Packaging Amber Glass Bottle
Assay Percent Range 97%
Chemical Name or Material 3-Bromo-2-methylimidazo[1, 2-a]pyridine
CAS 4805-70-3