Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-methylbenzotrifluoride, 97%

$551.47 - $551.47

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-methylbenzotrifluoride
CAS 69902-83-6
Molecular Formula C8H6BrF3
Formula Weight 239.04
MDL Number MFCD04038231
Synonym 2-Bromo-6-(trifluoromethyl)toluene; 2-Methyl-3-(trifluoromethyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2624906

  SDS
alfa aesar™
H2624906
5g Each for $551.47
Description & Specifications

Specifications

Formula Weight 239.04
Chemical Name or Material 3-Bromo-2-methylbenzotrifluoride
Molecular Formula C8H6BrF3
MDL Number MFCD04038231
CAS 69902-83-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Health Hazard 1 Warning
Boiling Point 35°C (1mmHg)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Synonym 2-Bromo-6-(trifluoromethyl)toluene; 2-Methyl-3-(trifluoromethyl)bromobenzene

Applications
3-Bromo-2-methylbenzotrifluoride is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.