Please login to your online account to display your discounted pricing

3-Bromo-2-methylbenzoic acid, 97%, ACROS Organics™

$169.45 - $169.45

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-methylbenzoic acid
CAS 76006-33-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H7BrO2
Formula Weight 215.05
MDL Number MFCD00270097
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC397300010

 
thermo fisher scientific
AC397300010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC397300050

 
thermo fisher scientific
AC397300050
Glass bottle 5g Each for $169.45
Description & Specifications

Specifications

Formula Weight 215.05
Physical Form Powder
Assay ≥96.0%
Quantity 1g
Color White to orange or brown
Melting Point 152° to 156°C
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
Molecular Formula C8H7BrO2
Assay Percent Range 97%
Chemical Name or Material 3-Bromo-2-methylbenzoic acid
CAS 76006-33-2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
MDL Number MFCD00270097
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.