missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-methylbenzoic acid, 97%

Manufacturer:  Alfa Aesar™ H5543606

 View more versions of this product

Catalog No. AAH5543606



Description

Description

Applications
3-Bromo-2-methylbenzoic Acid is a disubstituted benzoic acid used in the preparation of various biologically active compounds such as α-2 adrenoceptor agonists, Smoothened receptor anatogonists and HIV-1 entry inhibitors.

Solubility
Soluble in chloroform, methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
Specifications

Specifications

3-Bromo-2-methylbenzoic acid
5g
C8H7BrO2
2-bromo-6-carboxytoluene, 2-methyl-3-bromobenzoic acid, 3-bromo-2-methybenzoic acid, 3-bromo-2-methyl-benzoic acid, 3-bromo-2-methylbenzoicacid, 3-bromo-o-toluic acid, benzoic acid, 3-bromo-2-methyl, bromo-methylbenzoic acid, ksc377a4p, pubchem4717
BJGKVCKGUBYULR-UHFFFAOYSA-N
3-bromo-2-methylbenzoic acid
2735590
97%
152°C to 156°C
76006-33-2
MFCD00270097
Soluble in chloroform,methanol
CC1=C(C=CC=C1Br)C(=O)O
215.046
215.05

Chemical Identifiers

76006-33-2
215.046
BJGKVCKGUBYULR-UHFFFAOYSA-N
2735590
CC1=C(C=CC=C1Br)C(=O)O
C8H7BrO2
MFCD00270097
2-bromo-6-carboxytoluene, 2-methyl-3-bromobenzoic acid, 3-bromo-2-methybenzoic acid, 3-bromo-2-methyl-benzoic acid, 3-bromo-2-methylbenzoicacid, 3-bromo-o-toluic acid, benzoic acid, 3-bromo-2-methyl, bromo-methylbenzoic acid, ksc377a4p, pubchem4717
3-bromo-2-methylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents