Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-methoxypyridine, 95%

$397.78 - $923.65

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-methoxypyridine
CAS 13472-59-8
Molecular Formula C6H6BrNO
Formula Weight 188.02
MDL Number MFCD03095349
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2727903

  SDS
alfa aesar™
H2727903
1g Each for $397.78

AAH2727906

  SDS
alfa aesar™
H2727906
5g Each for $923.65
Description & Specifications

Specifications

Formula Weight 188.02
Assay 95%
Quantity 1g
Health Hazard 1 Warning
Chemical Name or Material 3-Bromo-2-methoxypyridine
CAS 13472-59-8
Flash Point 93°C (199°F)
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Molecular Formula C6H6BrNO
MDL Number MFCD03095349

Applications
It is employed in the facile synthesis of 2-unsubstituted benzofuran-3-carboxylates using diazo(trimethylsilyl)methyl magnesium bromide.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.