Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine, 98%

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine
CAS 124432-63-9
Formula Weight 256.02
MDL Number MFCD09972202
Beilstein 13639286
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6171806

  SDS
alfa aesar™
H6171806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6171814

  SDS
alfa aesar™
H6171814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 256.02
Chemical Name or Material 3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine
Quantity 5g
Physical Form Powder
Assay 98%
Color White
MDL Number MFCD09972202
CAS 124432-63-9
Beilstein 13639286
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.