missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine, 98%

Manufacturer:  Alfa Aesar™ H6171814

 View more versions of this product

Catalog No. AAH6171814



Specifications

Specifications

3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine
25g
C7H5BrF3NO
13639286
YUDYUCKEWCVXTE-UHFFFAOYSA-N
3-bromo-2-methoxy-5-(trifluoromethyl)pyridine
14837018
Powder
White
124432-63-9
MFCD09972202
3-bromo-2-methoxy-5-trifluoromethyl pyridine, bromides, acmc-209as0, pyridine,3-bromo-2-methoxy-5-trifluoromethyl, 2-methoxy-3-bromo-5-trifluoromethylpyridine, 3-bromo-5-trifluoromethyl-2-methoxypyridine
COC1=C(C=C(C=N1)C(F)(F)F)Br
256.022
256.02
98%

Chemical Identifiers

124432-63-9
256.022
YUDYUCKEWCVXTE-UHFFFAOYSA-N
14837018
COC1=C(C=C(C=N1)C(F)(F)F)Br
C7H5BrF3NO
MFCD09972202
3-bromo-2-methoxy-5-trifluoromethyl pyridine, bromides, acmc-209as0, pyridine,3-bromo-2-methoxy-5-trifluoromethyl, 2-methoxy-3-bromo-5-trifluoromethylpyridine, 3-bromo-5-trifluoromethyl-2-methoxypyridine
3-bromo-2-methoxy-5-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents