missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-2-hydroxypyridine, TCI America™

$261.25 - $907.52

Chemical Identifiers

CAS 13466-43-8
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 173.997
MDL Number MFCD03411569
InChI Key YDUGVOUXNSWQSW-UHFFFAOYSA-N
Synonym 3-bromo-2-hydroxypyridine, 3-bromopyridin-2-ol, 3-bromopyridin-2 1h-one, 3-bromo-2-pyridinol, 3-bromo-2 1h-pyridinone, 2 1h-pyridinone, 3-bromo, 2-hydroxy-3-bromopyridine, 3-bromo-2 1h-pyridone, bromopyridone, 3-bromopyridone
PubChem CID 818549
IUPAC Name 3-bromo-1H-pyridin-2-one
SMILES C1=CNC(=O)C(=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B23305G View Documents TCI America
B23305G
5g Each for $261.25
 
B233025G View Documents TCI America
B233025G
25g Each for $907.52
 
Specifications

Chemical Identifiers

13466-43-8
173.997
YDUGVOUXNSWQSW-UHFFFAOYSA-N
818549
C1=CNC(=O)C(=C1)Br
C5H4BrNO
MFCD03411569
3-bromo-2-hydroxypyridine, 3-bromopyridin-2-ol, 3-bromopyridin-2 1h-one, 3-bromo-2-pyridinol, 3-bromo-2 1h-pyridinone, 2 1h-pyridinone, 3-bromo, 2-hydroxy-3-bromopyridine, 3-bromo-2 1h-pyridone, bromopyridone, 3-bromopyridone
3-bromo-1H-pyridin-2-one

Specifications

Undesignated
5g
C5H4BrNO
3-bromo-2-hydroxypyridine, 3-bromopyridin-2-ol, 3-bromopyridin-2 1h-one, 3-bromo-2-pyridinol, 3-bromo-2 1h-pyridinone, 2 1h-pyridinone, 3-bromo, 2-hydroxy-3-bromopyridine, 3-bromo-2 1h-pyridone, bromopyridone, 3-bromopyridone
C1=CNC(=O)C(=C1)Br
173.997
174.00
≥97.0% (GC)
182°C
13466-43-8
MFCD03411569
YDUGVOUXNSWQSW-UHFFFAOYSA-N
3-bromo-1H-pyridin-2-one
818549
Crystal-Powder at 20°C
3-Bromo-2-hydroxypyridine
Safety and Handling

Safety and Handling

TSCA : No