Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-hydroxypyridine, 97%

$82.12 - $1,047.20

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-hydroxypyridine
CAS 13466-43-8
Molecular Formula C5H4BrNO
Formula Weight 174
MDL Number MFCD03411569
Beilstein 109846
Synonym 3-Bromo-2-pyridinol; 3-Bromo-2(1H)-pyridone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2001403

  SDS
alfa aesar™
L2001403
1g Each for $82.12

AAL2001406

  SDS
alfa aesar™
L2001406
5g Each for $274.23

AAL2001414

  SDS
alfa aesar™
L2001414
25g Each for $1,047.20
Description & Specifications

Specifications

Formula Weight 174
Assay 97%
CAS 13466-43-8
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
MDL Number MFCD03411569
Health Hazard 1 Warning
Beilstein 109846
Molecular Formula C5H4BrNO
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Melting Point 185° to 189°C
Synonym 3-Bromo-2-pyridinol; 3-Bromo-2(1H)-pyridone
Chemical Name or Material 3-Bromo-2-hydroxypyridine

Applications
3-Bromo-2-hydroxypyridine is sued as an active ingredient pharmaceuticals, in medicine.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.