Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-hydroxy-5-nitropyridine, 98%

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-hydroxy-5-nitropyridine
CAS 15862-33-6
Formula Weight 218.99
MDL Number MFCD03840431
Beilstein 135472
Synonym 3-Bromo-5-nitro-2-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6673906

 
alfa aesar™
H6673906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6673914

 
alfa aesar™
H6673914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 213° to 218°C
Formula Weight 218.99
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Beilstein 135472
Health Hazard 1 Danger
Chemical Name or Material 3-Bromo-2-hydroxy-5-nitropyridine
Physical Form Solid
Quantity 5g
MDL Number MFCD03840431
CAS 15862-33-6
Synonym 3-Bromo-5-nitro-2-pyridinol
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.