Please login to your online account to display your discounted pricing

3-bromo-2-hydroxy-5-nitrobenzaldehyde, Maybridge

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-hydroxy-5-nitrobenzaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4BrNO4
Formula Weight 246.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB03755DE

  SDS
fisher scientific company
BTB03755DE
Amber glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB03755DA

  SDS
fisher scientific company
BTB03755DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 246.02
Quantity 5g
Assay 97%
Assay Percent Range 97%
Chemical Name or Material 3-Bromo-2-hydroxy-5-nitrobenzaldehyde
Packaging Amber glass bottle
Molecular Formula C7H4BrNO4
CAS 16789-84-7