Please login to your online account to display your discounted pricing

3-bromo-2-hydroxy-5-nitrobenzaldehyde, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  BTB03755DA

 View more versions of this product

Catalog No. BTB03755DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 246.02
Quantity 1g
Assay 97%
CAS 16789-84-7
Assay Percent Range 97%
Chemical Name or Material 3-Bromo-2-hydroxy-5-nitrobenzaldehyde
Molecular Formula C7H4BrNO4

Specifications

Chemical Name or Material 3-Bromo-2-hydroxy-5-nitrobenzaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4BrNO4
Formula Weight 246.02