Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-hydroxy-5-methylpyridine, 97%

$73.97 - $259.16

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-hydroxy-5-methylpyridine
CAS 17282-02-9
Molecular Formula C6H6BrNO
Formula Weight 188.02
MDL Number MFCD03791312
Beilstein 1423374
Synonym 5-Bromo-2-hydroxy-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1984903

  SDS
alfa aesar™
L1984903
1g Each for $73.97

AAL1984906

  SDS
alfa aesar™
L1984906
5g Each for $259.16
Description & Specifications

Specifications

Chemical Name or Material 3-Bromo-2-hydroxy-5-methylpyridine
CAS 17282-02-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C6H6BrNO
Formula Weight 188.02
MDL Number MFCD03791312
Melting Point 156° to 158°C
Beilstein 1423374
Synonym 5-Bromo-2-hydroxy-3-picoline
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.