Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-fluorotoluene, 98%

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-fluorotoluene
CAS 59907-12-9
Formula Weight 189.02
MDL Number MFCD04972694
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6149214

 
alfa aesar™
H6149214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6149222

 
alfa aesar™
H6149222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 189.02
Melting Point 118° to 123°C
Assay 98%
Chemical Name or Material 3-Bromo-2-fluorotoluene
MDL Number MFCD04972694
CAS 59907-12-9
Refractive Index 1.5330
Quantity 25g
Density 1.503g/mL
Physical Form Liquid