missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-2-fluorobenzoic Acid, TCI America™

$90.14 - $271.95

Chemical Identifiers

CAS 161957-56-8
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD00665763
InChI Key UVKURTLVTLRSSM-UHFFFAOYSA-N
Synonym 3-bromo-2-fluoro-benzoic acid, 3-bromo-2-fluorobenzoicacid, 2-fluoro-3-bromobenzoic acid, benzoic acid, 3-bromo-2-fluoro, 3-bromo-2-fluoro benzoic acid, buttpark 14\01-30, 3-bromo-2-fluorobenzenecarboxylic acid, pubchem1318, acmc-209dnk, intermediates-zcf02220
PubChem CID 2736313
IUPAC Name 3-bromo-2-fluorobenzoic acid
SMILES C1=CC(=C(C(=C1)Br)F)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B39291G View Documents TCI America
B39291G
1g Each for $90.14
 
B39295G View Documents TCI America
B39295G
5g Each for $271.95
 
Specifications

Chemical Identifiers

161957-56-8
219.009
UVKURTLVTLRSSM-UHFFFAOYSA-N
2736313
C1=CC(=C(C(=C1)Br)F)C(=O)O
C7H4BrFO2
MFCD00665763
3-bromo-2-fluoro-benzoic acid, 3-bromo-2-fluorobenzoicacid, 2-fluoro-3-bromobenzoic acid, benzoic acid, 3-bromo-2-fluoro, 3-bromo-2-fluoro benzoic acid, buttpark 14\01-30, 3-bromo-2-fluorobenzenecarboxylic acid, pubchem1318, acmc-209dnk, intermediates-zcf02220
3-bromo-2-fluorobenzoic acid

Specifications

White-Yellow
1g
C7H4BrFO2
3-bromo-2-fluoro-benzoic acid, 3-bromo-2-fluorobenzoicacid, 2-fluoro-3-bromobenzoic acid, benzoic acid, 3-bromo-2-fluoro, 3-bromo-2-fluoro benzoic acid, buttpark 14\01-30, 3-bromo-2-fluorobenzenecarboxylic acid, pubchem1318, acmc-209dnk, intermediates-zcf02220
C1=CC(=C(C(=C1)Br)F)C(=O)O
219.009
219.01
≥98.0% (GC,T)
170°C
161957-56-8
MFCD00665763
UVKURTLVTLRSSM-UHFFFAOYSA-N
3-bromo-2-fluorobenzoic acid
2736313
Crystal-Powder at 20°C
3-Bromo-2-fluorobenzoic Acid
Safety and Handling

Safety and Handling

TSCA : No