Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-fluorobenzoic acid, 97%

$144.20 - $629.82

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-fluorobenzoic acid
CAS 161957-56-8
Molecular Formula C7H4BrFO2
Formula Weight 219.01
MDL Number MFCD00665763
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5023503

  SDS
alfa aesar™
H5023503
1g Each for $144.20

AAH5023506

  SDS
alfa aesar™
H5023506
5g Each for $629.82
Description & Specifications

Specifications

Formula Weight 219.01
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C7H4BrFO2
CAS 161957-56-8
Chemical Name or Material 3-Bromo-2-fluorobenzoic acid
Melting Point 168° to 170°C
Assay 97%
Health Hazard 1 Warning
MDL Number MFCD00665763
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.