Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-fluorobenzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-fluorobenzaldehyde
CAS 149947-15-9
Formula Weight 203.01
MDL Number MFCD09027089
Beilstein 13904499
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6465006

  SDS
alfa aesar™
H6465006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6465014

  SDS
alfa aesar™
H6465014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6465022

  SDS
alfa aesar™
H6465022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 43° to 46°C
Formula Weight 203.01
Chemical Name or Material 3-Bromo-2-fluorobenzaldehyde
Color Off-white
Physical Form Powder
Assay 97%
Quantity 5g
Sensitivity Light Sensitive
MDL Number MFCD09027089
CAS 149947-15-9
Beilstein 13904499
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C

Applications
It can be used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C