missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-fluoro-5-nitropyridine, 97%

$66.62 - $1,327.77

Chemical Identifiers

CAS 1868-58-2
Molecular Formula C5H2BrFN2O2
Molecular Weight (g/mol) 220.985
MDL Number MFCD08703668
InChI Key NLLPMLSUMJHTCJ-UHFFFAOYSA-N
Synonym 2-fluoro-3-bromo-5-nitropyridine, acmc-1celf, 3-bromanyl-2-fluoranyl-5-nitro-pyridine
PubChem CID 345124
IUPAC Name 3-bromo-2-fluoro-5-nitropyridine
SMILES C1=C(C=NC(=C1Br)F)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6150003 View Documents Alfa Aesar™
H6150003
1g Each for $66.62
 
AAH6150006 View Documents Alfa Aesar™
H6150006
5g Each for $296.08
 
AAH6150014 View Documents Alfa Aesar™
H6150014
25g Each for $1,327.77
 
Description

Description

Applications
3-Bromo-2-fluoro-5-nitropyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1868-58-2
220.985
NLLPMLSUMJHTCJ-UHFFFAOYSA-N
345124
C1=C(C=NC(=C1Br)F)[N+](=O)[O-]
C5H2BrFN2O2
MFCD08703668
2-fluoro-3-bromo-5-nitropyridine, acmc-1celf, 3-bromanyl-2-fluoranyl-5-nitro-pyridine
3-bromo-2-fluoro-5-nitropyridine

Specifications

1g
C5H2BrFN2O2
2-fluoro-3-bromo-5-nitropyridine, acmc-1celf, 3-bromanyl-2-fluoranyl-5-nitro-pyridine
C1=C(C=NC(=C1Br)F)[N+](=O)[O-]
220.985
220.98
3-Bromo-2-fluoro-5-nitropyridine
1868-58-2
MFCD08703668
NLLPMLSUMJHTCJ-UHFFFAOYSA-N
3-bromo-2-fluoro-5-nitropyridine
345124
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger