Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-cyanobenzeneboronic acid, 96%

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-cyanobenzeneboronic acid
CAS 1032231-32-5
Formula Weight 225.84
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6187103

  SDS
alfa aesar™
H6187103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61871MD

  SDS
alfa aesar™
H61871MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.84
Chemical Name or Material 3-Bromo-2-cyanobenzeneboronic acid
Physical Form Solid
Assay 96%
CAS 1032231-32-5
Quantity 1g