Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-chlorothiophene, 97%

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-chlorothiophene
CAS 40032-73-3
Molecular Formula C4H2BrClS
Formula Weight 197.48
MDL Number MFCD00043883
UN Number UN2810
Beilstein 1205539
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1931803

  SDS
alfa aesar™
L1931803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1931806

  SDS
alfa aesar™
L1931806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.48
CAS 40032-73-3
MDL Number MFCD00043883
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Bromo-2-chlorothiophene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Flash Point 85°C (185°F)
Molecular Formula C4H2BrClS
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Beilstein 1205539
Boiling Point 70°C (9mmHg)
UN Number UN2810
Assay 97%

Applications
3-Bromo-2-chlorothiophene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.