Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-chlorobenzotrifluoride, 97%

$192.41 - $580.31

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-chlorobenzotrifluoride
CAS 56131-47-6
Molecular Formula C7H3BrClF3
Formula Weight 259.44
MDL Number MFCD04115994
Synonym 2-Chloro-3-(trifluoromethyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2670003

 
alfa aesar™
H2670003
1g Each for $192.41

AAH2670006

 
alfa aesar™
H2670006
5g Each for $580.31
Description & Specifications

Specifications

Formula Weight 259.44
Chemical Name or Material 3-Bromo-2-chlorobenzotrifluoride
Assay 97%
Molecular Formula C7H3BrClF3
MDL Number MFCD04115994
CAS 56131-47-6
Synonym 2-Chloro-3-(trifluoromethyl)bromobenzene
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.