missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-chloro-6-methylpyridine, 98%

$29.15 - $524.32

Chemical Identifiers

CAS 185017-72-5
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD03095218
InChI Key JVDQYSIJBRTRMS-UHFFFAOYSA-N
Synonym 3-bromo-2-chloro-6-picoline, 5-bromo-6-chloro-2-picoline, 2-chloro-3-bromo-6-methylpyridine, 5-bromo-6-chloro-2-methylpyridine, pyridine, 3-bromo-2-chloro-6-methyl, 3-bromo-2-chloro-6-methyl-pyridine, pubchem1286, acmc-209emg, ksc538q8t, 3-bromo-2-chloro-6 picoline
PubChem CID 2734418
IUPAC Name 3-bromo-2-chloro-6-methylpyridine
SMILES CC1=NC(=C(C=C1)Br)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6415303 View Documents Alfa Aesar™
H6415303
1g Each for $29.15
 
AAH6415306 View Documents Alfa Aesar™
H6415306
5g Each for $113.20
 
AAH6415314 View Documents Alfa Aesar™
H6415314
25g Each for $524.32
 
Specifications

Chemical Identifiers

185017-72-5
206.467
JVDQYSIJBRTRMS-UHFFFAOYSA-N
2734418
CC1=NC(=C(C=C1)Br)Cl
C6H5BrClN
MFCD03095218
3-bromo-2-chloro-6-picoline, 5-bromo-6-chloro-2-picoline, 2-chloro-3-bromo-6-methylpyridine, 5-bromo-6-chloro-2-methylpyridine, pyridine, 3-bromo-2-chloro-6-methyl, 3-bromo-2-chloro-6-methyl-pyridine, pubchem1286, acmc-209emg, ksc538q8t, 3-bromo-2-chloro-6 picoline
3-bromo-2-chloro-6-methylpyridine

Specifications

White
185017-72-5
MFCD03095218
7757196
JVDQYSIJBRTRMS-UHFFFAOYSA-N
3-bromo-2-chloro-6-methylpyridine
2734418
Solid
3-Bromo-2-chloro-6-methylpyridine
1g
C6H5BrClN
UN2811
3-bromo-2-chloro-6-picoline, 5-bromo-6-chloro-2-picoline, 2-chloro-3-bromo-6-methylpyridine, 5-bromo-6-chloro-2-methylpyridine, pyridine, 3-bromo-2-chloro-6-methyl, 3-bromo-2-chloro-6-methyl-pyridine, pubchem1286, acmc-209emg, ksc538q8t, 3-bromo-2-chloro-6 picoline
CC1=NC(=C(C=C1)Br)Cl
206.467
206.47
98%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.