Learn More
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
$41.68 - $134.51
Chemical Identifiers
CAS | 71701-92-3 |
---|---|
Molecular Formula | C6H2BrClF3N |
Molecular Weight (g/mol) | 260.438 |
MDL Number | MFCD09878432 |
InChI Key | GVQLMGPWTHAUPV-UHFFFAOYSA-N |
Synonym | 3-bromo-2-chloro-5-trifluoromethyl pyridine, 2-chloro-3-bromo-5-trifluoromethylpyridine, abbypharma ap-13-5058, pyridine, 3-bromo-2-chloro-5-trifluoromethyl, pubchem15158, acmc-1bcir, ksc496i9b, 2-morpholin-4-ylethanesulfonicacid,hydrate, 2-chloro-3-bromo-5-trifluoromethyl pyridine |
PubChem CID | 13536061 |
IUPAC Name | 3-bromo-2-chloro-5-(trifluoromethyl)pyridine |
SMILES | C1=C(C=NC(=C1Br)Cl)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6443503
|
Thermo Scientific Chemicals
H6443503 |
1 g |
Each for $41.68
|
|
|||||
AAH6443506
|
Thermo Scientific Chemicals
H6443506 |
5 g |
Each for $134.51
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
71701-92-3 | |
260.438 | |
GVQLMGPWTHAUPV-UHFFFAOYSA-N | |
13536061 | |
C1=C(C=NC(=C1Br)Cl)C(F)(F)F |
C6H2BrClF3N | |
MFCD09878432 | |
3-bromo-2-chloro-5-trifluoromethyl pyridine, 2-chloro-3-bromo-5-trifluoromethylpyridine, abbypharma ap-13-5058, pyridine, 3-bromo-2-chloro-5-trifluoromethyl, pubchem15158, acmc-1bcir, ksc496i9b, 2-morpholin-4-ylethanesulfonicacid,hydrate, 2-chloro-3-bromo-5-trifluoromethyl pyridine | |
3-bromo-2-chloro-5-(trifluoromethyl)pyridine |
Specifications
71701-92-3 | |
Brown to Yellow | |
C6H2BrClF3N | |
1 g | |
3-bromo-2-chloro-5-trifluoromethyl pyridine, 2-chloro-3-bromo-5-trifluoromethylpyridine, abbypharma ap-13-5058, pyridine, 3-bromo-2-chloro-5-trifluoromethyl, pubchem15158, acmc-1bcir, ksc496i9b, 2-morpholin-4-ylethanesulfonicacid,hydrate, 2-chloro-3-bromo-5-trifluoromethyl pyridine | |
C1=C(C=NC(=C1Br)Cl)C(F)(F)F | |
260.438 | |
260.44 | |
Solid |
28°C to 32°C | |
98°C (208°F) | |
MFCD09878432 | |
UN2811 | |
GVQLMGPWTHAUPV-UHFFFAOYSA-N | |
3-bromo-2-chloro-5-(trifluoromethyl)pyridine | |
13536061 | |
97% | |
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures