missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2-butanone, 97%, stab. with <1% magnesium oxide

Manufacturer:  Alfa Aesar™ A1018618

 View more versions of this product

Catalog No. AAA1018618



Description

Description

Applications
3-Bromo-2-butanone is involved in the preparation of o-isopropyl S-3-oxobutan-2-yl dithiocarbonate by reaction with potassium o-isopropylxanthate, which on treatment with sulfuric acid gives 4,5-dimethyl-1,3-dithiol-2-one .

Solubility
Miscible with dichloromethane.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications

Specifications

3-Bromo-2-butanone
Pungent
50g
814-75-5
MFCD00013538
906773
BNBOUFHCTIFWHN-UHFFFAOYSA-N
3-bromobutan-2-one
13142
97%
>100°C (212°F)
69°C to 70°C (70mmHg)
Stabilized with <1% magnesium oxide
C4H7BrO
UN2922
3-bromo-2-butanone, 2-butanone, 3-bromo, 2-bromo-3-butanone, 3-bromo-butan-2-one, 1-bromoethyl methyl ketone, 2-bromobutan-2-one, 3-bromobutanone, 3-bromo-2-butanon, 2-bromo-3-oxobutane, 3-bromo-butan-2-on
CC(C(=O)C)Br
151.003
151.01

Chemical Identifiers

814-75-5
151.003
BNBOUFHCTIFWHN-UHFFFAOYSA-N
13142
CC(C(=O)C)Br
C4H7BrO
MFCD00013538
3-bromo-2-butanone, 2-butanone, 3-bromo, 2-bromo-3-butanone, 3-bromo-butan-2-one, 1-bromoethyl methyl ketone, 2-bromobutan-2-one, 3-bromobutanone, 3-bromo-2-butanon, 2-bromo-3-oxobutane, 3-bromo-butan-2-on
3-bromobutan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H314-H318-H312
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : EL7005000

TSCA : TSCA

SDS Documents