Please login to your online account to display your discounted pricing

3-Bromo-2-(bromomethyl)propionic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Bromo-2-(bromomethyl)propionic acid
CAS 41459-42-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96%
Assay Percent Range 97%
Molecular Formula C4H6Br2O2
Linear Formula BrCH2CH(CH2Br)CO2H
Formula Weight 245.9
MDL Number MFCD00010643
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC373190010

  SDS
thermo fisher scientific
AC373190010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96%
Formula Weight 245.9
Physical Form Crystalline Powder
Quantity 1g
Color Off-white to brown
Melting Point 98° to 103°C
CAS Max % 100.0
CAS Min % 96.0
Linear Formula BrCH2CH(CH2Br)CO2H
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Molecular Formula C4H6Br2O2
MDL Number MFCD00010643
Assay Percent Range 97%
CAS 41459-42-1
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material 3-Bromo-2-(bromomethyl)propionic acid
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.