Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2,6-dimethoxypyridine, 98%

$142.24 - $403.81

Chemical Identifiers

Chemical Name or Material 3-Bromo-2,6-dimethoxypyridine
CAS 13445-16-4
Molecular Formula C7H8BrNO2
Formula Weight 218.05
MDL Number MFCD08669938
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2756406

  SDS
alfa aesar™
H2756406
5g Each for $142.24

AAH2756414

  SDS
alfa aesar™
H2756414
25g Each for $403.81
Description & Specifications

Specifications

Formula Weight 218.05
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 13445-16-4
Assay 98%
Chemical Name or Material 3-Bromo-2,6-dimethoxypyridine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Molecular Formula C7H8BrNO2
Health Hazard 1 Warning
MDL Number MFCD08669938
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.