Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-2,6-dimethoxybenzoic acid, 98%

$57.09 - $184.72

Chemical Identifiers

Chemical Name or Material 3-Bromo-2,6-dimethoxybenzoic acid
CAS 73219-89-3
Molecular Formula C9H9BrO4
Formula Weight 261.07
MDL Number MFCD00052933
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1597103

 
alfa aesar™
A1597103
1g Each for $57.09

AAA1597106

 
alfa aesar™
A1597106
5g Each for $184.72
Description & Specifications

Specifications

Formula Weight 261.07
Chemical Name or Material 3-Bromo-2,6-dimethoxybenzoic acid
Assay 98%
Molecular Formula C9H9BrO4
MDL Number MFCD00052933
CAS 73219-89-3
Melting Point 144° to 148°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.