missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-2,6-difluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2621314

 View more versions of this product

Catalog No. AAH2621314



Description

Description

3-Bromo-2,6-difluorobenzaldehyde is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2,6-difluorobenzaldehyde is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
Specifications

Specifications

3-Bromo-2,6-difluorobenzaldehyde
53°C to 57°C
MFCD08282773
Air Sensitive
OXBHKEYDKAWFLS-UHFFFAOYSA-N
3-bromo-2,6-difluorobenzaldehyde
22139701
98%
398456-82-1
C7H3BrF2O
25 g
3-bromo-2,6-difluoro-benzaldehyde, benzaldehyde, 3-bromo-2,6-difluoro, acmc-209j8a, 3-bromo-2, 6-difluorobenzaldehyde, benzaldehyde,3-bromo-2,6-difluoro
C1=CC(=C(C(=C1F)C=O)F)Br
221.001
221

Chemical Identifiers

398456-82-1
221.001
OXBHKEYDKAWFLS-UHFFFAOYSA-N
3-bromo-2,6-difluorobenzaldehyde
C7H3BrF2O
MFCD08282773
22139701
C1=CC(=C(C(=C1F)C=O)F)Br
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only