Learn More
3-Bromo-2,6-difluorobenzaldehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2621306
Description
3-Bromo-2,6-difluorobenzaldehyde is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromo-2,6-difluorobenzaldehyde is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
Specifications
3-Bromo-2,6-difluorobenzaldehyde | |
53°C to 57°C | |
MFCD08282773 | |
Air Sensitive | |
OXBHKEYDKAWFLS-UHFFFAOYSA-N | |
3-bromo-2,6-difluorobenzaldehyde | |
22139701 | |
98% |
398456-82-1 | |
C7H3BrF2O | |
5 g | |
3-bromo-2,6-difluoro-benzaldehyde, benzaldehyde, 3-bromo-2,6-difluoro, acmc-209j8a, 3-bromo-2, 6-difluorobenzaldehyde, benzaldehyde,3-bromo-2,6-difluoro | |
C1=CC(=C(C(=C1F)C=O)F)Br | |
221.001 | |
221 |
Chemical Identifiers
398456-82-1 | |
221.001 | |
OXBHKEYDKAWFLS-UHFFFAOYSA-N | |
3-bromo-2,6-difluorobenzaldehyde |
C7H3BrF2O | |
MFCD08282773 | |
22139701 | |
C1=CC(=C(C(=C1F)C=O)F)Br |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only