missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-2,5-dichlorothiophene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1060306

 View more versions of this product

Catalog No. AAA1060306



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromo-2,5-dichlorothiophene
95°C to 97°C (14 mmHg)
1.608
5 g
thiophene,3-bromo-2,5-dichloro, zlchem 1326, acmc-209mjs, 2,5-dichloro bromo thiophene, 2,5-dichloro-3-bromothiophene, 3-bromo-2,5-dichloro thiophene, thiophene, 3-bromo-2,5-dichloro
ClC1=CC(Br)=C(Cl)S1
231.92
231.93
60404-18-4
C4HBrCl2S
MFCD00041440
1306272
PBUHOXBSIQJRNO-UHFFFAOYSA-N
3-bromo-2,5-dichlorothiophene
521925
97%

Chemical Identifiers

60404-18-4
231.92
PBUHOXBSIQJRNO-UHFFFAOYSA-N
3-bromo-2,5-dichlorothiophene
C4HBrCl2S
MFCD00041440
521925
ClC1=CC(Br)=C(Cl)S1
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only