missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-2,4-dimethyl-5-phenylthiophene 98.0+%, TCI America™
$162.59 - $486.08
Chemical Identifiers
CAS | 362513-28-8 |
---|---|
Molecular Formula | C12H11BrS |
Molecular Weight (g/mol) | 267.184 |
InChI Key | VJMJMLJTIYJVQY-UHFFFAOYSA-N |
PubChem CID | 44629772 |
IUPAC Name | 3-bromo-2,4-dimethyl-5-phenylthiophene |
SMILES | CC1=C(SC(=C1Br)C)C2=CC=CC=C2 |
Chemical Identifiers
362513-28-8 | |
267.184 | |
44629772 | |
CC1=C(SC(=C1Br)C)C2=CC=CC=C2 |
C12H11BrS | |
VJMJMLJTIYJVQY-UHFFFAOYSA-N | |
3-bromo-2,4-dimethyl-5-phenylthiophene |
Specifications
362513-28-8 | |
1 g | |
CC1=C(SC(=C1Br)C)C2=CC=CC=C2 | |
267.184 | |
267.18 | |
Crystals |
C12H11BrS | |
VJMJMLJTIYJVQY-UHFFFAOYSA-N | |
3-bromo-2,4-dimethyl-5-phenylthiophene | |
44629772 | |
≥98.0% (GC) | |
3-Bromo-2,4-dimethyl-5-phenylthiophene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator